Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ccdc65 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_153518.1 | NP_705738.1 | 5 | 947 | ||
GenBank mRNA | AF454432.1 | - | 5 | 947 | ||
AK077174.1 | - | 5 | 981 | |||
BC048073.1 | - | 5 | 967 | 6312435 |