Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign In
Don't have an account ? Create Account
  • Applications
    • Real-Time PCR
    • Oligos, Primers, Probes and Genes
    • Cloning
    • Protein Biology
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Cell Analysis
    • Mass Spectrometry
    • Chromatography
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Online Order
    • Custom Primers & TaqMan Probes
    • miRNA Mimics & Inhibitors
    • Stealth RNAi / siRNA
    • Silencer Select siRNAs
    • Custom DNA Oligos
    • GeneArt Services
    • GeneArt Strings DNA Fragments
    • TrueGuide CRISPR gRNA
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all online orderable products
  • Services
    • Custom Services
    • Instrument Qualification Services
    • Technical Services
    • Enterprise Services
    • Pipette Services
    • Sample Request
    • Instrument Maintenance Services
    • Repairs and Relocation Services
    • OEM and Licensing Services
    • Events, Seminars, Training
    • See all services
  • Support
    • Catalogs
    • Application Notes
    • Safety Data Sheets
    • Legal and Regulatory
    • Certificates of Analysis Search
    • Nunc/Nalgene Product Certificates
    • e-learning
    • FAQs
    • Learning Centers
    • Technical Support Centers
    • See all help and support topics
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign In
            Sign In
            Don't have an account ? Create Account
            • Connect Your Lab
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other BPNT1 SIRNA Assays ›
          Gene Symbol:
          BPNT1
          Gene Name:
          3'(2'), 5'-bisphosphate nucleotidase 1
          Gene Aliases:
          HEL20, PIP
          Chromosome Location:
          Chr.1: 220057482 - 220089916 on Build GRCh38
          UniGene ID:
          Hs.406134
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID 17361

          Purification
          Size
          Availability Inventoried
          Catalog # AM16708
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: BPNT1
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001286149.1 NP_001273078.1 4, 5 438
          NM_001286150.1 NP_001273079.1 4, 5 458
          NM_001286151.1 NP_001273080.1 4, 5 480
          NM_006085.5 NP_006076.4 5, 6 566
          XM_005272998.2 XP_005273055.1 5, 6 537
          XM_005272999.4 XP_005273056.1 5, 6 646
          XM_005273000.4 XP_005273057.1 5, 6 615
          XM_005273004.2 XP_005273061.1 5, 6 537
          XM_005273005.4 XP_005273062.1 5, 6 649
          XM_006711114.3 XP_006711177.1 4, 5 510
          XM_011509063.2 XP_011507365.1 4, 5 484
          XM_011509064.2 XP_011507366.1 3, 4 393
          XM_011509065.2 XP_011507367.1 5, 6 537
          XM_017000043.1 XP_016855532.1 4, 5 530
          XM_017000044.1 XP_016855533.1 3, 4 365
          XM_017000045.1 XP_016855534.1 5, 6 537
          XM_017000046.1 XP_016855535.1 4, 5 487
          GenBank mRNA AF125042.1 - 5, 6 503
          AJ249339.1 - 5, 6 531
          AK291943.1 - 5, 6 501
          AK298476.1 - 4, 5 383
          AK300777.1 - 4, 5 403
          AK315962.1 - 4, 5 460
          BC017801.1 - 5, 6 505 4338207
          BG428446.1 - 5, 6 556
          EU794594.1 - 5, 6 503

          Back To Top

          More Information


          Other Species Targeted

          This siRNA also targets the following transcripts in Rat: NM_171990, XM_006250430, XM_006250431, XM_006250429, AJ000347, and others.
          This siRNA also targets the following transcripts in Mouse: NM_011794, AK008091, AK154435, AF125043, AK150877, and others.

          Panther Classification:

          Molecular Function -

          phosphatase

          Gene Ontology Categories:

          Function(s) Process(es)

          nucleobase-containing compound metabolic process
          nervous system development
          dephosphorylation
          phosphatidylinositol phosphorylation
          3'-phosphoadenosine 5'-phosphosulfate metabolic process
          magnesium ion binding
          inositol-1,4-bisphosphate 1-phosphatase activity
          3'(2'),5'-bisphosphate nucleotidase activity

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Online Orderable Products
          • Privacy Policy for Online Ordering
          • Act on Specified Commercial Transactions
          • About Returns
          • About Displayed Prices
          • About Shipping Fees
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Study information disclosure
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Seminars
          • Blog
          About Thermo Fisher Plus Icon Minus Icon
          • Corporate Information
          • Social Responsibility (CSR)
          • Enhancing Customer Experience
          • Careers Careers
          • News
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Japan flag icon
          Japan

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-s4sd5:80/100.66.78.82:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0