Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Afap1l1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_178928.4 | NP_849259.2 | 12 | 1401 | ||
GenBank mRNA | AK035227.1 | - | 12 | 1401 | ||
AK053714.1 | - | 11 | 1636 | |||
BC094505.1 | - | 9 | 839 | |||
BC138923.1 | - | 12 | 1362 | 8861945 |