siRNA Details
Gene Symbol: Hmgb2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008252.3 | NP_032278.1 | 3 | 392 | ||
GenBank mRNA | AK003773.1 | - | 3 | 298 | ||
AK008443.1 | - | 3 | 303 | |||
AK011467.1 | - | 3 | 299 | |||
AK012568.1 | - | 3 | 359 | |||
AK076075.1 | - | 3 | 328 | |||
AK135296.1 | - | 3 | 298 | |||
AK135297.1 | - | 3 | 298 | |||
AK146212.1 | - | 3 | 327 | |||
AK151164.1 | - | 3 | 306 | |||
AK153858.1 | - | 3 | 301 | |||
BC002050.1 | - | 3 | 272 | 3489780 | ||
BC046759.1 | - | 3 | 296 | 5693598 | ||
BC083108.1 | - | 3 | 290 | 5320702 | ||
Z46757.1 | - | 3 | 301 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Panther Classification:
