Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mtmr14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026849.2 | NP_081125.2 | 5 | 627 | ||
GenBank mRNA | AK034867.1 | - | 5 | 574 | ||
AK178962.1 | - | 3 | 126 | |||
AK181484.1 | - | 3 | 118 | |||
AK189716.1 | - | 3 | 119 | |||
AK191101.1 | - | 4 | 249 | |||
AK207311.1 | - | 2 | 74 | |||
AK210125.1 | - | 4 | 249 | |||
AK218803.1 | - | 4 | 249 | |||
BC012436.1 | - | 3 | 155 | |||
BC018294.1 | - | 5 | 577 | 4021243 | ||
CD805346.1 | - | 3 | 173 |