Search
Search

Hnrnpm| Gene Symbol: Hnrnpm | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001109913.1 | NP_001103383.1 | 8, 9 | 736 | ||
| NM_029804.3 | NP_084080.1 | 7, 8 | 853 | |||
| GenBank mRNA | AK011521.1 | - | 7, 8 | 799 | ||
| AK133911.1 | - | 7, 8 | 788 | |||
| AK145343.1 | - | 8, 9 | 674 | |||
| AK159848.1 | - | 8, 9 | 668 | |||
| AK166670.1 | - | 7, 8 | 788 | |||
| AK168347.1 | - | 8, 9 | 671 | |||
| AK169101.1 | - | 8, 9 | 671 | |||
| AK171892.1 | - | 8, 9 | 671 | |||
| AK220299.1 | - | 8, 9 | 696 | |||
| BC017539.1 | - | 8, 9 | 666 | |||
| BC035525.1 | - | 8, 9 | 671 | |||
| BC054785.1 | - | 2255 | ||||
| BC065172.1 | - | 8, 9 | 666 | 6415170 | ||