siRNA Details
Gene Symbol: ACAA2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006111.2 | NP_006102.2 | 3 | 617 | ||
GenBank mRNA | AK054673.1 | - | 3 | 617 | ||
AK225457.1 | - | 3 | 268 | |||
AK314462.1 | - | 3 | 619 | |||
BC001918.1 | - | 3 | 239 | 2820744 | ||
D16294.1 | - | 3 | 268 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Panther Classification:
