Search
Search

Pofut2| Gene Symbol: Pofut2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_030262.3 | NP_084538.2 | 3 | 436 | ||
| GenBank mRNA | AF455270.1 | - | 3 | 426 | ||
| AK132373.1 | - | 3 | 433 | |||
| AK147414.1 | - | 2 | 2031 | |||
| AK148244.1 | - | 3 | 435 | |||
| BC018194.1 | - | 3 | 434 | |||
| BC147077.1 | - | 3 | 503 | 9056064 | ||
| BC147078.1 | - | 3 | 503 | 9056063 | ||
| CO814561.1 | - | 436 | ||||