Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Cell Analysis
Antibodies
Mass Spectrometry
Cell Culture
Laboratory Instruments
Clinical and Diagnostics
Chromatography
Laboratory Equipment
Laboratory Supplies
Molecular Biology and Nucleic Acid Analysis
Sequence-Specific Nucleic Acid Products
See all product categories
Applications
Cell Culture and Transfection
Flow Cytometry
Cancer Research
Chromatography
Sequencing
PCR
Lab Solutions
Allergy Diagnostics
See all applications and techniques
Services
Instrument Services
Cell Biology Services
Custom Services
Training Services
Enterprise Services
Lab Informatics Services
Financial and Leasing Services
Partnering and Licensing Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
See all services
Help and Support
How to Order
Promotions and Online Offers
Contact Us
Change Location
Create a New Account
See all help and support topics
Popular
Our Instagram
Our Facebook
Blog Behind the Bench
Customer Experience Center (CEC)
Ecommerce Exclusives
Special Offers
Contact Us
Quick Order
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Contact Us
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Custom Products & Projects
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other UCN3 SIRNA Assays ›
Gene Symbol:
Ucn3
Gene Name:
urocortin 3
Gene Aliases:
-
Chromosome Location:
Chr.13: 3940688 - 3945349 on Build GRCm38
UniGene ID:
Mm.313662
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID
175082
Purification
Standard
HPLC
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
286.00
Your Price
Online offer:
277.65
277.65
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: Ucn3
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_031250.5
NP_112540.2
2
750
GenBank mRNA
AK148451.1
-
2
751
BC119433.1
-
1
235
40041175
BC119434.1
-
1
232
Back To Top
More Information
Gene Ontology Categories:
Function(s)
Process(es)
response to stress
adenylate cyclase-activating G-protein coupled receptor signaling pathway
neuropeptide signaling pathway
digestion
cellular response to nutrient levels
positive regulation of insulin secretion
positive regulation of membrane potential
G-protein coupled receptor binding
hormone activity
neuropeptide hormone activity
protein binding
corticotropin-releasing hormone receptor 2 binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-blue-55ff9ddd88-lffdw:80/100.66.79.31:80.
git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.42.0-2026.01.03-1.0