Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Enterprise Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other NEDD4L SIRNA Assays ›
          Gene Symbol:
          Nedd4l
          Gene Name:
          neural precursor cell expressed, developmentally down-regulated gene 4-like
          Gene Aliases:
          1300012C07Rik, Nedd4-2, Nedd4b
          Chromosome Location:
          Chr.18: 64887756 - 65217826 on Build GRCm38
          UniGene ID:
          Mm.98668
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          siRNA ID 175197

          Purification
          Size
          Availability Inventoried
          Catalog # AM16708
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: Nedd4l
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001114386.1 NP_001107858.1 10, 11 1116
          NM_031881.2 NP_114087.2 9, 10 1165
          GenBank mRNA AF277232.1 - 9, 10 798
          AK004969.1 - 9, 10 858
          AK031842.1 - 9, 10 846
          AK042621.1 - 10, 11 799
          AK122283.1 - 9, 10 968
          AK163497.1 - 8, 9 646
          AK197688.1 - 1, 2 27
          BC039746.1 - 9, 10 809 3157137
          BC071210.1 - 9, 10 786 4972426
          BC082281.1 - 9, 10 835

          Back To Top

          More Information


          Other Species Targeted

          This siRNA also targets the following transcripts in Human: NM_001243960, NM_015277, NM_001144968, NM_001144967, NM_001144969, and others.
          This siRNA also targets the following transcripts in Rat: NM_001008300, GQ160815, GQ160816.

          Panther Classification:

          Molecular Function -

          ubiquitin-protein ligase

          Gene Ontology Categories:

          Function(s) Process(es)

          negative regulation of systemic arterial blood pressure
          regulation of membrane depolarization
          protein monoubiquitination
          response to salt stress
          response to metal ion
          positive regulation of sodium ion transport
          negative regulation of sodium ion transport
          protein ubiquitination
          regulation of ion transmembrane transport
          regulation of membrane potential
          protein ubiquitination involved in ubiquitin-dependent protein catabolic process
          proteasome-mediated ubiquitin-dependent protein catabolic process
          ubiquitin-dependent protein catabolic process via the multivesicular body sorting pathway
          positive regulation of protein catabolic process
          regulation of membrane repolarization
          protein K48-linked ubiquitination
          ventricular cardiac muscle cell action potential
          regulation of potassium ion transmembrane transporter activity
          negative regulation of potassium ion transmembrane transporter activity
          negative regulation of potassium ion transmembrane transport
          negative regulation of sodium ion transmembrane transport
          positive regulation of dendrite extension
          negative regulation of protein localization to cell surface
          negative regulation of sodium ion transmembrane transporter activity
          regulation of bicellular tight junction assembly
          positive regulation of cation channel activity
          positive regulation of caveolin-mediated endocytosis
          ubiquitin-protein transferase activity
          protein binding
          UDP-N-acetylmuramoylalanyl-D-glutamyl-2,6-diaminopimelate-D-alanyl-D-alanine ligase activity
          potassium channel regulator activity
          ligase activity
          sodium channel regulator activity
          ribosomal S6-glutamic acid ligase activity
          potassium channel inhibitor activity
          sodium channel inhibitor activity
          coenzyme F420-0 gamma-glutamyl ligase activity
          coenzyme F420-2 alpha-glutamyl ligase activity
          ion channel binding
          ubiquitin protein ligase activity
          protein-glycine ligase activity
          protein-glycine ligase activity, initiating
          protein-glycine ligase activity, elongating
          tubulin-glycine ligase activity
          protein-glutamic acid ligase activity
          tubulin-glutamic acid ligase activity

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-db7ps:80/100.66.74.176:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0