Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Enterprise Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other PDE4D SIRNA Assays ›
          Gene Symbol:
          PDE4D
          Gene Name:
          phosphodiesterase 4D
          Gene Aliases:
          ACRDYS2, DPDE3, HSPDE4D, PDE43, PDE4DN2, STRK1
          Chromosome Location:
          Chr.5: 58969038 - 60488098 on Build GRCh38
          UniGene ID:
          Hs.117545
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID 17522
          Validated

          Purification
          Size
          Availability Inventoried
          Catalog # AM51331
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: PDE4D
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001104631.1 NP_001098101.1 8 1242
          NM_001165899.1 NP_001159371.1 10 1133
          NM_001197218.1 NP_001184147.1 8 981
          NM_001197219.1 NP_001184148.1 8 759
          NM_001197220.1 NP_001184149.1 8 1397
          NM_001197221.1 NP_001184150.1 4 717
          NM_001197222.1 NP_001184151.1 3 803
          NM_001197223.1 NP_001184152.1 3 593
          NM_006203.4 NP_006194.2 8 785
          XM_005248537.3 XP_005248594.1 8 1939
          XM_005248538.4 XP_005248595.1 9 3840
          XM_011543469.1 XP_011541771.1 11 1222
          XM_011543470.2 XP_011541772.1 11 1146
          XM_011543471.1 XP_011541773.1 10 1099
          XM_011543473.1 XP_011541775.1 10 1047
          XM_011543477.2 XP_011541779.1 10 1049
          XM_017009565.1 XP_016865054.1 11 1259
          XM_017009566.1 XP_016865055.1 11 1149
          XM_017009567.1 XP_016865056.1 9 887
          XM_017009568.1 XP_016865057.1 11 1133
          XM_017009569.1 XP_016865058.1 10 1012
          XM_017009570.1 XP_016865059.1 7 1832
          XM_017009571.1 XP_016865060.1 10 1055
          GenBank mRNA AF012073.1 - 8 995
          AF012074.1 - 4 731
          AF536975.1 - 3 400
          AF536976.1 - 10 956
          AF536977.1 - 8 759
          AJ250852.1 - 4 327
          AJ250854.1 - 9 922
          AY245866.1 - 10 1098
          AY245867.1 - 8 1397
          BC036319.1 - 3 595 4828512
          L20970.1 - 8 793
          U02882.1 - 7 563
          U50157.1 - 3 401
          U50158.1 - 4 315
          U50159.1 - 8 736
          U79571.1 - 3 806

          Back To Top

          More Information


          Matching Taqman Gene Expression Assay:


          Hs00174810_m1

          Panther Classification:

          Molecular Function -

          phosphodiesterase

          Gene Ontology Categories:

          Function(s) Process(es)

          regulation of heart rate
          cAMP catabolic process
          smooth muscle contraction
          aging
          regulation of receptor activity
          regulation of release of sequestered calcium ion into cytosol by sarcoplasmic reticulum
          cAMP-mediated signaling
          neutrophil chemotaxis
          regulation of cAMP metabolic process
          positive regulation of interferon-gamma production
          positive regulation of interleukin-2 production
          positive regulation of interleukin-5 production
          negative regulation of peptidyl-serine phosphorylation
          multicellular organism growth
          negative regulation of heart contraction
          T cell receptor signaling pathway
          regulation of ryanodine-sensitive calcium-release channel activity
          establishment of endothelial barrier
          cellular response to lipopolysaccharide
          cellular response to epinephrine stimulus
          adrenergic receptor signaling pathway
          regulation of cardiac muscle cell contraction
          adrenergic receptor signaling pathway involved in positive regulation of heart rate
          regulation of cell communication by electrical coupling involved in cardiac conduction
          negative regulation of relaxation of cardiac muscle
          3',5'-cyclic-nucleotide phosphodiesterase activity
          3',5'-cyclic-AMP phosphodiesterase activity
          protein binding
          drug binding
          enzyme binding
          cAMP binding
          beta-2 adrenergic receptor binding
          ion channel binding
          metal ion binding
          ATPase binding
          scaffold protein binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-lffdw:80/100.66.79.31:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0