Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cecr6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_033567.1 | NP_291045.1 | 1 | 1528 | ||
GenBank mRNA | AF277399.1 | - | 1 | 1528 | ||
AK140695.1 | - | 2 | 598 | |||
BC137862.1 | - | 1 | 1528 | 8860881 |