Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PNLIPRP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001303135.1 | NP_001290064.1 | 4 | 333 | ||
NM_006229.3 | NP_006220.1 | 4 | 343 | |||
XM_011539868.1 | XP_011538170.1 | 4 | 476 | |||
XM_011539869.1 | XP_011538171.1 | 4 | 330 | |||
GenBank mRNA | AK130236.1 | - | 4 | 331 | ||
AK307063.1 | - | 4 | 330 | |||
BC005233.1 | - | 4 | 343 | |||
BC025784.2 | - | 3 | 306 | 5226231 | ||
BP323340.1 | - | 4 | 333 | |||
CR749299.1 | - | 4 | 342 | |||
CR749596.1 | - | 4 | 344 | |||
M93283.1 | - | 4 | 325 |