Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ddx50 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053183.2 | NP_444413.1 | 2 | 324 | ||
GenBank mRNA | AF334104.1 | - | 2 | 325 | ||
AK041281.1 | - | 2 | 223 | |||
AK162423.1 | - | 3 | 442 | |||
BC055440.1 | - | 2 | 318 | |||
BC075714.1 | - | 2 | 276 | |||
BC090996.1 | - | 2 | 344 | 6757408 |