Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
CDMO and Clinical Trial Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Instrument Management
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other CSF2RA SIRNA Assays ›
Gene Symbol:
CSF2RA
Gene Name:
colony stimulating factor 2 receptor alpha subunit
Gene Aliases:
CD116, CDw116, CSF2R, CSF2RAX, CSF2RAY, CSF2RX, CSF2RY, GM-CSF-R-alpha, GMCSFR, GMR, SMDP4
Chromosome Location:
Chr.X: 1268800 - 1325097 on Build GRCh38
UniGene ID:
Hs.520937
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
17582
Purification
HPLC
Standard
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: CSF2RA
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001161529.1
NP_001155001.1
6
601
NM_001161530.1
NP_001155002.1
3
305
NM_001161531.1
NP_001155003.1
5
473
NM_001161532.1
NP_001155004.1
3
242
NM_006140.4
NP_006131.2
5
473
NM_172245.2
NP_758448.1
5
449
NM_172246.2
NP_758449.1
5
473
NM_172247.2
NP_758450.1
3
305
NM_172249.2
NP_758452.1
5
473
NR_027760.1
5
473
XM_011546165.2
XP_011544467.1
4
351
XM_011546166.1
XP_011544468.1
4
351
XM_011546167.2
XP_011544469.1
4
351
XM_011546168.2
XP_011544470.1
4
351
XM_011546169.1
XP_011544471.1
4
351
XM_011546170.2
XP_011544472.1
4
351
XM_011546173.1
XP_011544475.1
4
351
XM_011546174.2
XP_011544476.1
4
351
XM_011546175.2
XP_011544477.1
4
351
XM_017029287.1
XP_016884776.1
5
463
XM_017029288.1
XP_016884777.1
5
440
GenBank mRNA
AK293086.1
-
5
431
AK301395.1
-
3
224
AK309630.1
-
5
434
AL833597.1
-
5
410
BC002635.1
-
5
449
3606186
BC071835.1
-
6
571
4650755
BI868829.1
-
5
448
DQ841258.1
-
3
279
L29348.1
-
5
428
L29349.1
-
5
428
M64445.1
-
5
441
M73832.1
-
3
297
U93096.1
-
5
428
X17648.1
-
5
428
X54935.1
-
3
279
Back To Top
More Information
Panther Classification:
Molecular Function -
transmembrane signal receptor
Gene Ontology Categories:
Function(s)
Process(es)
MAPK cascade
peptidyl-tyrosine phosphorylation
granulocyte colony-stimulating factor signaling pathway
positive regulation of GTPase activity
cellular protein metabolic process
cellular response to granulocyte macrophage colony-stimulating factor stimulus
protein tyrosine kinase activity
receptor activity
granulocyte colony-stimulating factor receptor activity
Ras guanyl-nucleotide exchange factor activity
protein binding
granulocyte colony-stimulating factor binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-55874ff697-twsbf:80/10.239.167.23:80.
git-commit: a2f5e26549390d543f5d2d1fbad8092df65fbda4
git-url: git@github.com:thermofisher/magellan-core.git
git-branch: origin/release/2.10.0-2024.06.23-1.0