Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ano10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001271873.1 | NP_001258802.1 | 4 | 551 | ||
NM_133979.3 | NP_598740.1 | 4 | 551 | |||
GenBank mRNA | AK043011.1 | - | 4 | 514 | ||
AK046091.1 | - | 4 | 503 | |||
AK079622.1 | - | 4 | 502 | |||
AK080947.1 | - | 4 | 503 | |||
AK138414.1 | - | 4 | 537 | |||
AK195077.1 | - | 2 | 126 | |||
BC099688.1 | - | 4 | 474 | 6313494 |