Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: OAS3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006187.3 | NP_006178.2 | 16 | 4947 | ||
XM_005253889.3 | XP_005253946.1 | 16 | 4874 | |||
XM_017019363.1 | XP_016874852.1 | 16 | 4721 | |||
GenBank mRNA | AB044545.1 | - | 16 | 5028 | ||
AF063613.1 | - | 16 | 4883 | |||
BG765554.1 | - | 1 | 69 |