siRNA Details
Gene Symbol: Mgat4b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145926.3 | NP_666038.3 | 9, 10 | 1342 | ||
GenBank mRNA | AB053218.1 | - | 9, 10 | 1044 | ||
AK155336.1 | - | 9, 10 | 1343 | |||
AK192721.1 | - | 1, 2 | 65 | |||
BC026638.1 | - | 4, 5 | 450 | 4984907 | ||
BC031613.1 | - | 3, 4 | 320 | 4216720 | ||
CX205743.1 | - | 6, 7 | 602 | |||
CX206051.1 | - | 1, 2 | 101 |
