Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Oaf | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_178644.3 | NP_848759.1 | 4 | 869 | ||
GenBank mRNA | AK051349.1 | - | 4 | 869 | ||
AK150279.1 | - | 4 | 856 | |||
BC025514.1 | - | 4 | 877 | 5309953 | ||
BC026022.1 | - | 4 | 833 | 5098241 | ||
BC028824.1 | - | 4 | 904 | 5097873 | ||
BC034836.1 | - | 4 | 835 | 5125217 |