Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tiparp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_178892.5 | NP_849223.2 | 2 | 545 | ||
GenBank mRNA | AK028529.1 | - | 2 | 546 | ||
AK032453.1 | - | 2 | 529 | |||
AK150248.1 | - | 2 | 527 | |||
AK162094.1 | - | 2 | 551 | |||
AK163347.1 | - | 2 | 584 | |||
BC068173.1 | - | 2 | 519 | 30616032 |