Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Aox3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_023617.2 | NP_076106.2 | 13 | 1295 | ||
GenBank mRNA | AF172276.2 | - | 13 | 1371 | ||
AK004700.1 | - | 2 | 91 | |||
AK030109.1 | - | 9 | 835 | |||
AK149434.1 | - | 13 | 1256 | |||
AY347569.1 | - | 13 | 1172 | |||
AY347570.1 | - | 13 | 1172 | |||
BC049853.1 | - | 13 | 1247 | |||
BC138876.1 | - | 13 | 1274 | 8861898 |