Search
Search

Tmem80| Gene Symbol: Tmem80 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001141950.1 | NP_001135422.1 | 6 | 891 | ||
| NM_027797.3 | NP_082073.1 | 6 | 960 | |||
| GenBank mRNA | AK017460.1 | - | 6 | 960 | ||
| AK049405.1 | - | 2 | 821 | |||
| AK134821.1 | - | 6 | 892 | |||
| AK169083.1 | - | 6 | 850 | |||
| BC022756.1 | - | 6 | 835 | 4922223 | ||