Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Osbpl3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001163645.1 | NP_001157117.1 | 12, 13 | 1783 | ||
NM_027881.3 | NP_082157.2 | 12, 13 | 1501 | |||
GenBank mRNA | AK004768.1 | - | 12, 13 | 1503 | ||
AK037114.1 | - | 12, 13 | 1470 | |||
AK173006.1 | - | 13, 14 | 4715 | |||
BC044866.1 | - | 11, 12 | 1428 | 5003631 | ||
BC086485.1 | - | 12, 13 | 1695 | 5344992 |