Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: R3hdm2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001168292.1 | NP_001161764.1 | 18 | 1983 | ||
NM_001168293.1 | NP_001161765.1 | 16 | 2202 | |||
NM_027900.4 | NP_082176.4 | 16 | 2163 | |||
GenBank mRNA | AK004884.1 | - | 6 | 743 | ||
AK043545.1 | - | 16 | 2164 | |||
AK122416.1 | - | 17 | 1841 | |||
AX775759.1 | - | 7 | 860 | |||
BC023826.1 | - | 2 | 86 | |||
BC043083.1 | - | 14 | 1541 | |||
BC053099.1 | - | 11 | 1246 | |||
BC064442.1 | - | 11 | 1214 | |||
BC082999.1 | - | 4 | 604 | |||
BC117927.2 | - | 16 | 1945 | 40097626 |