Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Who We Serve
Biotech
Biopharma
CDMO
Lab Diagnostics
Industrial and Applied Sciences
Promotions
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Promos
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other NSUN4 SIRNA Assays ›
Gene Symbol:
Nsun4
Gene Name:
NOL1/NOP2/Sun domain family, member 4
Gene Aliases:
2310010O12Rik, 2810405F18Rik, AI507900, Shtap
Chromosome Location:
Chr.4: 116031770 - 116053881 on Build GRCm38
UniGene ID:
Mm.282560
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID
177469
Purification
Standard
HPLC
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: Nsun4
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_028142.4
NP_082418.1
2
214
GenBank mRNA
AK012994.1
-
2
215
BC024628.1
-
2
134
4165638
FJ882982.1
-
2
2760
Back To Top
More Information
Panther Classification:
Molecular Function -
RNA methyltransferase
Gene Ontology Categories:
Function(s)
Process(es)
rRNA processing
rRNA methylation
methylation
ribosome biogenesis
mature ribosome assembly
positive regulation of mitochondrial translation
rRNA (adenine-N6,N6-)-dimethyltransferase activity
RNA binding
protein C-terminal carboxyl O-methyltransferase activity
tRNA (guanine-N2-)-methyltransferase activity
methyltransferase activity
C-methyltransferase activity
N-methyltransferase activity
O-methyltransferase activity
S-methyltransferase activity
RNA methyltransferase activity
mRNA methyltransferase activity
tRNA methyltransferase activity
protein methyltransferase activity
2-polyprenyl-6-methoxy-1,4-benzoquinone methyltransferase activity
rRNA methyltransferase activity
rRNA (uridine-2'-O-)-methyltransferase activity
rRNA (adenine-N6-)-methyltransferase activity
DNA-methyltransferase activity
tRNA (guanine-N1-)-methyltransferase activity
rRNA (cytosine-C5-)-methyltransferase activity
selenocysteine methyltransferase activity
arginine N-methyltransferase activity
protein-arginine N-methyltransferase activity
lysine N-methyltransferase activity
protein-lysine N-methyltransferase activity
tRNA (uracil) methyltransferase activity
tRNA (guanine) methyltransferase activity
tRNA (adenine) methyltransferase activity
tRNA (cytosine) methyltransferase activity
tRNA (cytosine-5-)-methyltransferase activity
tRNA (adenine-N1-)-methyltransferase activity
rRNA (adenine) methyltransferase activity
rRNA (cytosine) methyltransferase activity
rRNA (guanine) methyltransferase activity
rRNA (uridine) methyltransferase activity
transferase activity
protein C-terminal leucine carboxyl O-methyltransferase activity
1-phenanthrol methyltransferase activity
protein-arginine N5-methyltransferase activity
rRNA binding
methylarsonite methyltransferase activity
dimethylarsinite methyltransferase activity
4,5-dihydroxybenzo(a)pyrene methyltransferase activity
1-hydroxypyrene methyltransferase activity
1-hydroxy-6-methoxypyrene methyltransferase activity
demethylmenaquinone methyltransferase activity
cobalt-precorrin-6B C5-methyltransferase activity
cobalt-precorrin-7 C15-methyltransferase activity
cobalt-precorrin-5B C1-methyltransferase activity
cobalt-precorrin-3 C17-methyltransferase activity
dimethylamine methyltransferase activity
hydroxyneurosporene-O-methyltransferase activity
tRNA (adenine-57, 58-N(1)-) methyltransferase activity
methylamine-specific methylcobalamin:coenzyme M methyltransferase activity
trimethylamine methyltransferase activity
methanol-specific methylcobalamin:coenzyme M methyltransferase activity
monomethylamine methyltransferase activity
P-methyltransferase activity
Se-methyltransferase activity
2-phytyl-1,4-naphthoquinone methyltransferase activity
tRNA (uracil-2'-O-)-methyltransferase activity
tRNA (cytosine-2'-O-)-methyltransferase activity
phosphomethylethanolamine N-methyltransferase activity
tRNA (cytosine-3-)-methyltransferase activity
small ribosomal subunit rRNA binding
rRNA (cytosine-2'-O-)-methyltransferase activity
rRNA (cytosine-N4-)-methyltransferase activity
trihydroxyferuloyl spermidine O-methyltransferase activity
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-659f68c6f4-vnb2r:80/100.66.79.197:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0