Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Anks3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_028301.4 | NP_082577.2 | 3 | 470 | ||
GenBank mRNA | AK012494.1 | - | 3 | 419 | ||
AK089037.1 | - | 3 | 445 | |||
AK129471.1 | - | 3 | 414 | |||
AK182539.1 | - | 2 | 298 | |||
BC050929.1 | - | 3 | 406 | 6807194 | ||
BC051143.1 | - | 3 | 437 | 6531161 | ||
CX242757.1 | - | 3 | 433 |