Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Setd5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_028385.1 | NP_082661.1 | 3 | 311 | ||
GenBank mRNA | AK122551.1 | - | 3 | 311 | ||
AK164036.1 | - | 1 | 1062 | |||
BC056502.1 | - | 5 | 790 | |||
BC083184.1 | - | 2 | 135 | 30536831 |