Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Paqr8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_028829.3 | NP_083105.3 | 3 | 902 | ||
GenBank mRNA | AF313617.1 | - | 4 | 961 | ||
AK006107.2 | - | 3 | 888 | |||
AK029723.1 | - | 3 | 885 | |||
AK029772.1 | - | 3 | 919 | |||
AK031969.1 | - | 3 | 915 | |||
AK048045.1 | - | 3 | 917 | |||
AK147376.1 | - | 3 | 907 | |||
AY424297.1 | - | 3 | 917 | |||
BC059813.1 | - | 3 | 902 | 6843180 |
Set Membership: |
G-protein coupled receptor |