Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slx1b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_029420.4 | NP_083696.2 | 6 | 1696 | ||
GenBank mRNA | AK014753.1 | - | 5 | 1818 | ||
BC055909.1 | - | 6 | 1647 |