Search
Search

Gstk1| Gene Symbol: Gstk1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_029555.2 | NP_083831.1 | 5 | 493 | ||
| GenBank mRNA | AK002661.1 | - | 5 | 491 | ||
| AY279096.1 | - | 5 | 406 | |||
| BC038151.1 | - | 5 | 420 | |||
| BC051507.1 | - | 5 | 457 | |||
| BC058163.1 | - | 5 | 426 | 5100572 | ||