Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fam63a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_133858.4 | NP_598619.2 | 4 | 1554 | ||
NM_199475.1 | NP_955769.1 | 4 | 1554 | |||
GenBank mRNA | AB075603.1 | - | 3 | 565 | ||
AK036838.1 | - | 4 | 1546 | |||
AK087859.1 | - | 4 | 1367 | |||
AK162551.1 | - | 4 | 1274 | |||
AK169673.1 | - | 4 | 1554 | |||
AK208613.1 | - | 3 | 187 | |||
BC039762.1 | - | 3 | 372 | |||
BC051048.1 | - | 3 | 437 | |||
BC057901.1 | - | 2 | 83 | |||
BC094388.1 | - | 4 | 1371 | 5708548 | ||
BC132301.1 | - | 3 | 799 | 40130578 | ||
BC144749.1 | - | 3 | 799 | 9053297 |