Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pogk | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001142948.1 | NP_001136420.1 | 4 | 441 | ||
NM_175170.4 | NP_780379.2 | 5 | 775 | |||
GenBank mRNA | AK031716.1 | - | 5 | 728 | ||
AK048067.1 | - | 5 | 734 | |||
AK053898.1 | - | 5 | 752 | |||
AK122520.1 | - | 4 | 713 | |||
BC072587.1 | - | 395 | 6829796 |