Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc44a4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_023557.3 | NP_076046.1 | 19 | 1919 | ||
GenBank mRNA | AK008864.1 | - | 8 | 881 | ||
AK145621.1 | - | 19 | 1911 | |||
AK164684.1 | - | 18 | 2687 | |||
AK166620.1 | - | 16 | 1821 | |||
BC010808.1 | - | 19 | 1917 | 4240658 | ||
BC052652.1 | - | 19 | 1917 | 30034507 |