Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rffl | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001007465.3 | NP_001007466.1 | 7 | 1370 | ||
NM_001164569.1 | NP_001158041.1 | 8 | 1514 | |||
NM_001164570.1 | NP_001158042.1 | 8 | 1456 | |||
NM_001164571.1 | NP_001158043.1 | 6 | 1289 | |||
NM_026097.3 | NP_080373.1 | 6 | 1397 | |||
GenBank mRNA | AK006757.1 | - | 6 | 1385 | ||
AK007189.1 | - | 6 | 1385 | |||
AK028095.1 | - | 6 | 1386 | |||
AK128983.1 | - | 8 | 1457 | |||
AK150213.1 | - | 7 | 1303 | |||
AK150541.1 | - | 8 | 1476 | |||
AK159442.1 | - | 7 | 1329 | |||
AK169393.1 | - | 6 | 1230 | |||
AK169988.1 | - | 7 | 1373 | |||
BC118517.1 | - | 6 | 1172 | 40105713 |