Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc39a8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001135149.1 | NP_001128621.1 | 7 | 1245 | ||
NM_001135150.1 | NP_001128622.1 | 7 | 1395 | |||
NM_026228.5 | NP_080504.3 | 7 | 1354 | |||
GenBank mRNA | AK014895.1 | - | 7 | 1358 | ||
AK016853.1 | - | 7 | 1132 | |||
AK028215.1 | - | 7 | 1143 | |||
AK030918.1 | - | 1 | 61 | |||
AK089060.1 | - | 6 | 1254 | |||
BC006731.1 | - | 6 | 1056 | 3155670 |