Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zc3h6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_178404.3 | NP_848491.2 | 3 | 682 | ||
GenBank mRNA | AK014766.1 | - | 3 | 676 | ||
AK039171.1 | - | 3 | 677 | |||
BC058173.1 | - | 3 | 647 |