Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Samd8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026283.2 | NP_080559.1 | 5 | 1115 | ||
GenBank mRNA | AK006211.1 | - | 5 | 1116 | ||
AK015593.1 | - | 5 | 1093 | |||
AK075749.1 | - | 5 | 1066 | |||
AK147234.1 | - | 5 | 919 | |||
AK147528.1 | - | 5 | 919 | |||
BC100476.1 | - | 5 | 1130 | 30916247 | ||
BC116745.1 | - | 5 | 1103 | 40126064 | ||
BC116747.1 | - | 5 | 1103 | 40126066 |