Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Stx17 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026343.2 | NP_080619.2 | 4 | 374 | ||
GenBank mRNA | AK011400.1 | - | 4 | 368 | ||
AK014718.1 | - | 3 | 313 | |||
AK018526.1 | - | 4 | 364 | |||
AK034718.1 | - | 4 | 363 | |||
AK135240.1 | - | 4 | 370 | |||
AK154503.1 | - | 4 | 375 | |||
BC028639.1 | - | 4 | 361 | 1243532 |