Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Coq9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026452.2 | NP_080728.1 | 9 | 1021 | ||
GenBank mRNA | AK004527.1 | - | 5 | 446 | ||
AK009179.1 | - | 9 | 1033 | |||
AK154343.1 | - | 9 | 1046 | |||
AK158979.1 | - | 9 | 1040 | |||
BC036386.1 | - | 9 | 1021 | 1067021 |