Search
Search

Mak16| Gene Symbol: Mak16 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_026453.3 | NP_080729.1 | 6 | 473 | ||
| GenBank mRNA | AK015369.1 | - | 6 | 514 | ||
| AK075982.1 | - | 6 | 474 | |||
| AK075983.1 | - | 6 | 474 | |||
| BC013079.1 | - | 6 | 488 | 3499754 | ||