Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ube2f | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026454.3 | NP_080730.1 | 10 | 1104 | ||
GenBank mRNA | AK007549.1 | - | 10 | 1075 | ||
AK030594.1 | - | 1084 | ||||
AK153481.1 | - | 10 | 1105 | |||
BC006057.1 | - | 4 | 617 | |||
BC016117.1 | - | 11 | 1386 | 4486610 |