Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Srsf6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026499.4 | NP_080775.3 | 6 | 1853 | ||
GenBank mRNA | AK133872.1 | - | 6 | 1834 | ||
AK146293.1 | - | 6 | 1831 | |||
AK159519.1 | - | 6 | 1832 | |||
BC012039.1 | - | 6 | 1840 | 2648111 | ||
BC029312.1 | - | 1 | 946 |