Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mfsd10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026660.2 | NP_080936.1 | 13 | 1762 | ||
GenBank mRNA | AK003763.1 | - | 8 | 1030 | ||
AK018736.1 | - | 13 | 1775 | |||
AK089055.1 | - | 13 | 1779 | |||
AK144407.1 | - | 4 | 482 | |||
AK152438.1 | - | 13 | 1748 | |||
AK178371.1 | - | 1 | 100 | |||
AK191138.1 | - | 1 | 100 | |||
AK198393.1 | - | 1 | 100 | |||
AK199918.1 | - | 1 | 100 | |||
BC023012.1 | - | 13 | 1762 | 5376230 | ||
BC064775.1 | - | 13 | 1660 | 30023877 |