Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dcaf12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026893.3 | NP_081169.2 | 2 | 599 | ||
GenBank mRNA | AK005099.1 | - | 2 | 545 | ||
AK030173.1 | - | 2 | 551 | |||
AK031089.1 | - | 2 | 529 | |||
AK083369.1 | - | 1 | 1549 | |||
AK087517.1 | - | 2 | 482 | |||
AK090028.1 | - | 2 | 548 | |||
AK143635.1 | - | 2 | 552 | |||
AK173289.1 | - | 2 | 459 | |||
AK188223.1 | - | 1 | 107 | |||
BC023852.1 | - | 2 | 535 | 5346910 |