Search
Search

Vps36| Gene Symbol: Vps36 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_027338.1 | NP_081614.1 | 14 | 1237 | ||
| GenBank mRNA | AK005810.1 | - | 3 | 404 | ||
| AK150754.1 | - | 14 | 1258 | |||
| AK151035.1 | - | 14 | 1258 | |||
| AK156241.1 | - | 1252 | ||||
| AK161865.1 | - | 1289 | ||||
| BC010811.1 | - | 14 | 1237 | 4210911 | ||