Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rbm25 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_027349.3 | NP_081625.3 | 7, 8 | 930 | ||
GenBank mRNA | AK011403.1 | - | 7, 8 | 911 | ||
AK142132.1 | - | 1, 2 | 206 | |||
AK151910.1 | - | 7, 8 | 882 | |||
AK162135.1 | - | 1023 | ||||
AK164367.1 | - | 7, 8 | 895 | |||
BC010792.1 | - | 6, 7 | 676 | |||
BC023749.1 | - | 5, 6 | 639 | |||
BC158104.1 | - | 6, 7 | 810 | 9007388 |