Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rnf168 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_027355.2 | NP_081631.2 | 6 | 3863 | ||
GenBank mRNA | BC046815.1 | - | 6 | 3835 | 5702430 |