Search
Search

Tbc1d19| Gene Symbol: Tbc1d19 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_144517.4 | NP_653100.1 | 18, 19 | 1459 | ||
| GenBank mRNA | AK012836.1 | - | 18, 19 | 1410 | ||
| AK076128.1 | - | 18, 19 | 1407 | |||
| AK083084.1 | - | 18, 19 | 1430 | |||
| AK212437.1 | - | 1, 2 | 56 | |||
| BC020138.1 | - | 18, 19 | 1418 | 3983701 | ||