Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ncapd2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_146171.1 | NP_666283.1 | 11, 12 | 1436 | ||
GenBank mRNA | AK013380.1 | - | 2, 3 | 250 | ||
AK129068.1 | - | 11, 12 | 1401 | |||
AK156143.1 | - | 11, 12 | 1466 | |||
BC006673.1 | - | 4, 5 | 582 | |||
BC008592.1 | - | 2, 3 | 330 | |||
BC025460.1 | - | 11, 12 | 1456 | |||
BC026982.1 | - | 11, 12 | 1436 | 5364933 | ||
BC029133.1 | - | 11, 12 | 1393 | 4951596 |