Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mela | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK090120.1 | - | 838 | |||
AK090126.1 | - | 840 | ||||
AK090128.1 | - | 840 | ||||
U63133.1 | - | 834 |